MilliporeSigma
  • Functional characterization of the promoter region of human TNFAIP1 gene.
Functional characterization of the promoter region of human TNFAIP1 gene.
Molecular biology reports (2009-07-14)
Mingjun Liu, Zhenhua Sun, Aidong Zhou, Hong Li, Liping Yang, Chang Zhou, Rushi Liu, Xiang Hu, Jianlin Zhou, Shuanglin Xiang, Jian Zhang
ABSTRACT
Tumor necrosis factor, alpha-induced protein 1 (TNFAIP1) is an immediate-early response gene of endothelium induced by TNF alpha. However, little is really known concerning the TNFAIP1 expression regulation. To better understand how TNFAIP1 expression is regulated, we functionally characterized the promoter region of human TNFAIP1 gene. Deletion mutation analysis, gel electrophoretic mobility shift, and site-directed mutagenesis assays allowed the identification of one functional Sp1-binding site within the human TNFAIP1 core promoter region. Moreover, chromatin immunoprecipitation analysis indicated that Sp1 was associated in vivo with the TNFAIP1 promoter. Further, Sp1 overexpression enhanced TNFAIP1 promoter activity. These findings suggest that Sp1 is implicated in the control of basal TNFAIP1 gene expression. Accordingly, Sp1 is supposed to be involved in the elevation of TNFAIP1 in response to TNF alpha induction, and thus participate in inflammation-associated angiogenesis.