MilliporeSigma
  • Small molecules enhance CRISPR genome editing in pluripotent stem cells.
Small molecules enhance CRISPR genome editing in pluripotent stem cells.
Cell stem cell (2015-02-07)
Chen Yu, Yanxia Liu, Tianhua Ma, Kai Liu, Shaohua Xu, Yu Zhang, Honglei Liu, Marie La Russa, Min Xie, Sheng Ding, Lei S Qi
ABSTRACT
The bacterial CRISPR-Cas9 system has emerged as an effective tool for sequence-specific gene knockout through non-homologous end joining (NHEJ), but it remains inefficient for precise editing of genome sequences. Here we develop a reporter-based screening approach for high-throughput identification of chemical compounds that can modulate precise genome editing through homology-directed repair (HDR). Using our screening method, we have identified small molecules that can enhance CRISPR-mediated HDR efficiency, 3-fold for large fragment insertions and 9-fold for point mutations. Interestingly, we have also observed that a small molecule that inhibits HDR can enhance frame shift insertion and deletion (indel) mutations mediated by NHEJ. The identified small molecules function robustly in diverse cell types with minimal toxicity. The use of small molecules provides a simple and effective strategy to enhance precise genome engineering applications and facilitates the study of DNA repair mechanisms in mammalian cells.
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
3′-Azido-3′-deoxythymidine, ≥98% (HPLC)
Sigma-Aldrich
L755507, ≥98% (HPLC)