MilliporeSigma
  • Synthesis of thiol-linked neoglycopolymers and thermo-responsive glycomicelles as potential drug carrier.
Synthesis of thiol-linked neoglycopolymers and thermo-responsive glycomicelles as potential drug carrier.
Chemical communications (Cambridge, England) (2009-02-26)
Gaojian Chen, Sadik Amajjahe, Martina H Stenzel
ABSTRACT
Homopolymer and block copolymer bearing carbohydrate side chain functionality were obtained by grafting glucothiose onto alkene functional scaffolds via a thiol-ene click reaction and the resulting copolymer was used to form thermo-responsive micelles as a potential drug carrier.
Sigma-Aldrich
4-Pentenoic anhydride, 98%