MilliporeSigma
  • Characterization of a naturally occurring ErbB4 isoform that does not bind or activate phosphatidyl inositol 3-kinase.
Characterization of a naturally occurring ErbB4 isoform that does not bind or activate phosphatidyl inositol 3-kinase.
Oncogene (1999-06-03)
K Elenius, C J Choi, S Paul, E Santiestevan, E Nishi, M Klagsbrun
ABSTRACT
Receptor tyrosine kinases regulate cell behavior by activating specific signal transduction cascades. Epidermal growth factor (EGF) receptor tyrosine kinases include ErbB1, ErbB2, ErbB3 and ErbB4. ErbB4 is a tyrosine kinase receptor that binds neuregulins (NRG) and several other EGF family members. Reverse transcriptase polymerase chain reaction (RT-PCR) analysis identified two isoforms of ErbB4 that differed in their cytoplasmic domain sequences. Specifically, RT-PCR using primers flanking the putative phosphatidyl inositol 3-kinase (PI3-K) binding site of ErbB4 generated two specific bands when human and mouse heart and kidney tissues were analysed. Cloning and sequencing of these RT-PCR products revealed that one of the ErbB4 isoforms (ErbB4 CYT-2) lacked a 16 amino acid sequence including a putative PI3-K binding site, that was present in the other isoform (ErbB4 CYT-1). RT-PCR analysis of mouse tissues suggested that the expression of ErbB4 CYT-1 and ErbB4 CYT-2 was tissue-specific. Heart, breast and abdominal aorta expressed predominantly ErbB4 CYT-1 whereas neural tissues and kidney expressed predominantly ErbB4 CYT-2. To ascertain whether the absence of the putative PI3-K binding site in ErbB4 CYT-2 also resulted in the loss of PI3-K activity, NIH3T3 cell lines overexpressing ErbB4 CYT-1 or ErbB4 CYT-2 were produced. NRG-1 bound to and stimulated equivalent tyrosine phosphorylation of both isoforms. However, unlike ErbB4 CYT-1, the ErbB4 CYT-2 isoform was unable to bind the p85 subunit of PI3-K and to stimulate PI3-K activity in these cells. Furthermore, tyrosine phosphorylation of p85 or association of PI3-K activity with phosphotyrosine was not induced in NRG-1 treated cells expressing ErbB4 CYT-2, indicating that this isoform was incapable of activating PI3-K even indirectly. It was concluded that a novel naturally occurring ErbB4 isoform exists with a deletion of the cytoplasmic domain sequence required for the activation of the PI3-K intracellular signal transduction pathway and that this is the only PI3-K binding site in ErbB4.