MilliporeSigma
  • Polypeptide N-acetylgalactosaminyltransferase 2 regulates cellular metastasis-associated behavior in gastric cancer.
Polypeptide N-acetylgalactosaminyltransferase 2 regulates cellular metastasis-associated behavior in gastric cancer.
International journal of molecular medicine (2012-09-21)
Dong Hua, Li Shen, Lan Xu, Zhi Jiang, Yinghui Zhou, Aihuan Yue, Shitao Zou, Zhihong Cheng, Shiliang Wu
ABSTRACT
Aberrant glycosylation of cell surface glycoprotein due to specific alterations of glycosyltransferase activity is usually associated with invasion and metastasis of cancer, particularly of gastric carcinomas. Polypeptide N-acetylgalactosaminyltransferase 2 (ppGalNAc-T2), which catalyzes initiation of mucin-type O-glycosylation, is also involved in tumor migration and invasion. However, a comprehensive understanding of how ppGalNAc-T2 correlates with the metastasic potential of human gastric cancer is not currently available. In the present study, ppGalNAc-T2 was detected in a variety of human poorly differentiated tumor cells, and expression appeared to be higher in SGC7901 gastric cancer cells. In addition, we investigated the potential effects of ppGalNAc-T2 on growth and metastasis-associated behavior in SGC7901 cells after stable transfection with ppGalNAc-T2 sense and antisense vectors. We found that cell proliferation, adhesion and invasion were decreased in ppGalNAc-T2 overexpressed cells but increased in ppGalNAc-T2 downregulated cells. Therefore, we attempted to clarify the mechanisms underlying the anti-metastatic activities of ppGalNAc-T2. Further investigation indicated that overexpression of ppGalNAc-T2 is involved in the inhibition of matrix metalloproteinase (MMP)-2 expression at both the protein and mRNA levels, which may be associated with ppGalNAc-T2 suppressing the expression of transforming growth factor (TGF)-β1. However, it did not exhibit any apparent correlation with MMP-14 expression levels. Our data show the effect of ppGalNAc-T2 on proliferation, adhesion or invasion of SGC7901 gastric cancer cells, suggesting that ppGalNAc-T2 may exert anti-proliferative and anti-metastatic activity through the decrease of MMP-2 and TGF-β1. These results indicate that ppGalNAc‑T2 may be used as a novel therapeutic target for human gastric cancer treatment.