MilliporeSigma
  • Evaluation of circulating Transforming growth factor-beta1, Glypican-3 and Golgi protein-73 mRNAs expression as predictive markers for hepatocellular carcinoma in Egyptian patients.
Evaluation of circulating Transforming growth factor-beta1, Glypican-3 and Golgi protein-73 mRNAs expression as predictive markers for hepatocellular carcinoma in Egyptian patients.
Molecular biology reports (2013-11-05)
Gehan H Ibrahim, Moushira A Mahmoud, Nagwa M Aly
ABSTRACT
Hepatocellular carcinoma (HCC) incidence is fast-growing especially in countries highly prevalent with viral hepatitis. Its poor prognosis has driven the research toward the discovery of sensitive markers for early detection. We investigated the usefulness of serum Transforming growth factor-beta1 (TGF-β1), Glypican-3 (GPC3), and Golgi protein-73 (GP73) mRNAs as early biomarkers in HCC Egyptian patients chronically infected with hepatitis C virus (HCV) in comparison with serum alpha-fetoprotein (AFP). Using semi-quantitative RT-PCR and densitometry analysis, circulating TGF-β1, GPC3, and GP73 mRNAs expressions were estimated in 15 healthy adults, 15 chronic HCV (CHC) patients and 25 HCC patients. Serum GP73 expression percentage in HCC group was significantly higher than controls (100 vs. 40 %, P ≤ 0.001) and when compared to elevated serum AFP levels (100 vs. 36 %, P ≤ 0.001). TGF-β1 and GP73 expression means were also higher in HCC patients than controls and CHC patients (P < 0.05). GPC3 expression showed higher frequency in CHC patients compared to HCC group (80 vs. 28 %, P = 0.0016). According to the study cutoffs, serum TGF-β1 and GP73 mRNAs showed 60 and 96 % sensitivities for HCC diagnosis with 100 and 95 % specificities, respectively. Furthermore, elevated GP73 mRNA expression levels in early HCC were significantly increased compared to those of TGF-β1 mRNA and to high serum AFP (92.3 vs. 53.8 and 23.1 %; P = 0.03 and 0.0004, respectively). In conclusion, circulating TGF-β1 and GP73 mRNAs could be useful biomarkers for HCV-induced HCC diagnosis. Moreover, serum GP73 mRNA is sensitive for early cancer detection than AFP and TGF-β1 mRNA. However, these results need further validation studies.