MilliporeSigma
  • Aerobic degradation of tetrachloroethylene by toluene-o-xylene monooxygenase of Pseudomonas stutzeri OX1.
Aerobic degradation of tetrachloroethylene by toluene-o-xylene monooxygenase of Pseudomonas stutzeri OX1.
Nature biotechnology (2000-07-11)
D Ryoo, H Shim, K Canada, P Barbieri, T K Wood
ABSTRACT
Tetrachloroethylene (PCE) is thought to have no natural source, so it is one of the most difficult contaminants to degrade biologically. This common groundwater pollutant was thought completely nonbiodegradable in the presence of oxygen. Here we report that the wastewater bacterium Pseudomonas stutzeri OX1 degrades aerobically 0. 56 micromol of 2.0 micromol PCE in 21 h (Vmax approximately 2.5 nmol min(-1) mg(-1) protein and KM approximately 34 microM). These results were corroborated by the generation of 0.48 micromol of the degradation product, chloride ions. This degradation was confirmed to be a result of expression of toluene-o-xylene monooxygenase (ToMO) by P. stutzeri OX1, since cloning and expressing this enzyme in Escherichia coli led to the aerobic degradation of 0.19 micromol of 2.0 micromol PCE and the generation of stoichiometric amounts of chloride. In addition, PCE induces formation of ToMO, which leads to its own degradation in P. stutzeri OX1. Degradation intermediates reduce the growth rate of this strain by 27%.
Sigma-Aldrich
Tetrachloroethylene, anhydrous, ≥99%
Sigma-Aldrich
Tetrachloroethylene, ACS reagent, ≥99.0%