- Impact of genetic variation in SORCS1 on memory retention.
Impact of genetic variation in SORCS1 on memory retention.
We previously reported that genetic variants in SORCS1 increase the risk of AD, that over-expression of SorCS1 reduces γ-secretase activity and Aβ levels, and that SorCS1 suppression increases γ-secretase processing of APP and Aβ levels. We now explored the effect of variation in SORCS1 on memory. We explored associations between SORCS1-SNPs and memory retention in the NIA-LOAD case control dataset (162 cases,670 controls) and a cohort of Caribbean Hispanics (549 cases,544 controls) using single marker and haplotype analyses. Three SNPs in intron 1, were associated with memory retention in the NIA-LOAD dataset or the Caribbean Hispanic dataset (rs10884402(A allele:β = -0.15,p = 0.008), rs7078098(C allele:β = 0.18,p = 0.007) and rs950809(C allele:β = 0.17,p = 0.008)) and all three SNPs were significant in a meta-analysis of both datasets (0.002<p<0.03). The corresponding A-T-T haplotype for these SNPs was associated with lower scores in both datasets (p = 0.02,p = 0.0009), and the complementary G-C-C haplotype was associated with higher scores in NIA-LOAD (p = 0.02). These associations were restricted to cases. Variation in intron 1 in SORCS1 is associated with memory changes in AD.