MilliporeSigma
  • Kanosamine biosynthesis: a likely source of the aminoshikimate pathway's nitrogen atom.
Kanosamine biosynthesis: a likely source of the aminoshikimate pathway's nitrogen atom.
Journal of the American Chemical Society (2002-09-05)
Jiantao Guo, John W Frost
ABSTRACT
The biosynthetic source of the nitrogen atom incorporated into the aminoshikimate pathway has remained a question for some time. 3-Amino-3-deoxy-D-fructose 6-phosphate has previously been demonstrated to be a precursor to 4-amino-3,4-dideoxy-D-arabino-heptulosonic acid 7-phosphate and 3-amino-5-hydroxybenzoic acid via the inferred intermediacy of 1-deoxy-1-imino-D-erythrose 4-phosphate in Amycolatopsis mediterranei cell-free extract. This investigation examines the possibility that the natural product kanosamine might be a precursor to 3-amino-3-deoxy-D-fructose 6-phosphate. Kanosamine 6-phosphate was synthesized by a chemoenzymatic route and incubated in A. mediterranei cell-free lysate along with D-ribose 5-phosphate and phosphoenolpyruvate. Formation of 4-amino-3,4-dideoxy-D-arabino-heptulosonic acid 7-phosphate and 3-amino-5-hydroxybenzoic acid was observed. Subsequent incubation in A. mediterranei cell-free lysate of glutamine and NAD with UDP-glucose resulted in the formation of kanosamine. The bioconversion of UDP-glucose into kanosamine along with the bioconversion of kanosamine 6-phosphate into 4-amino-3,4-dideoxy-D-arabino-heptulosonic acid 7-phosphate and 3-amino-5-hydroxybenzoic acid suggests that kanosamine biosynthesis is the source of the aminoshikimate pathway's nitrogen atom.