MilliporeSigma
  • Use of a thick-film capillary column for the analysis of organic acids in body fluids.
Use of a thick-film capillary column for the analysis of organic acids in body fluids.
Journal of chromatography (1993-05-19)
W Meier-Augenstein, G F Hoffmann, B Holmes, J L Jones, W L Nyhan, L Sweetman
ABSTRACT
An improved method for the identification and quantification of organic acids in body fluids employing capillary gas chromatography-mass spectrometry has been developed. A thick-film capillary column, that combines the properties of a capillary column with those of a megabore column, has been successfully introduced into an existing method. Analysis over a concentration range from 1 mumol/l to 500 mumol/l body fluid is possible. This permits the assay of samples that are usually obtained in small volumes, e.g. cerebrospinal fluid.