MilliporeSigma
  • Knockdown of BAMBI inhibits β-catenin and transforming growth factor β to suppress metastasis of gastric cancer cells.
Knockdown of BAMBI inhibits β-catenin and transforming growth factor β to suppress metastasis of gastric cancer cells.
Molecular medicine reports (2014-06-11)
Kai Liu, Xilin Song, Heng Ma, Liqing Liu, Xiaowen Wen, Jianyong Yu, Longgang Wang, Sanyuan Hu
ABSTRACT
The upregulation of bone morphogenetic protein and activin membrane‑bound inhibitor (BAMBI) has been observed in several types of malignant cancer, including thyroid, ovarian, liver and colorectal cancer. However, the pathological role and the regulatory mechanism of BAMBI in gastric cancer remain to be elucidated. The present study revealed that the expression of BAMBI was upregulated in gastric cancer tissue, and was correlated with tumor metastasis, disease recurrence and low survival rates in patients. Knockdown of BAMBI in aggressive gastric cancer cell lines significantly inhibited their malignant behavior, including in vitro invasion and cell proliferation. β‑catenin expression was downregulated as a result of knocking down of BAMBI, and TGF-β was downregulated in a similar manner. These results demonstrated the association between BAMBI expression and gastric cancer progression, and indicate a promising direction for developing novel strategies to improve the prognosis and therapy of gastric cancer.
Sigma-Aldrich
Anti-TGF β1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-BAMBI antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Human BAMBI ELISA, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Anti-β-Catenin antibody produced in rabbit, whole antiserum