MilliporeSigma
  • Synthesis of pyrryl aryl sulfones targeted at the HIV-1 reverse transcriptase.
Synthesis of pyrryl aryl sulfones targeted at the HIV-1 reverse transcriptase.
Archiv der Pharmazie (1995-03-01)
M Artico, R Silvestri, G Stefancich, S Massa, E Pagnozzi, D Musu, F Scintu, E Pinna, E Tinti, P La Colla
ABSTRACT
Various aryl 1-pyrryl sulfones were synthesized and tested as inhibitors of HIV-1. 2-Nitrophenyl-2-ethoxycarbonyl-1-pyrryl sulfone, the most active among test derivatives, was selected as lead compound of the aryl pyrryl sulfone series. The in vitro anti-HIV-1 activity and cytotoxicity of 41 compounds is reported. Some structure-activity relationships are discussed also in comparison with the known NPPS (2-nitrophenyl phenyl sulfone).