MilliporeSigma
  • Synthesis of chiral sulfoximine-based thioureas and their application in asymmetric organocatalysis.
Synthesis of chiral sulfoximine-based thioureas and their application in asymmetric organocatalysis.
Beilstein journal of organic chemistry (2012-09-29)
Marcus Frings, Isabelle Thomé, Carsten Bolm
ABSTRACT
For the first time, chiral sulfoximine derivatives have been applied as asymmetric organocatalysts. In combination with a thiourea-type backbone the sulfonimidoyl moiety leads to organocatalysts showing good reactivity in the catalytic desymmetrization of a cyclic meso-anhydride and moderate enantioselectivity in the catalytic asymmetric Biginelli reaction. Straightforward synthetic routes provide the newly designed thiourea-sulfoximine catalysts in high overall yields without affecting the stereohomogeneity of the sulfur-containing core fragment.
Sigma-Aldrich
3,5-Bis(trifluoromethyl)phenyl isothiocyanate, 98%