MilliporeSigma
  • Benzylamine histamine H(3) antagonists and serotonin reuptake inhibitors.
Benzylamine histamine H(3) antagonists and serotonin reuptake inhibitors.
Bioorganic & medicinal chemistry letters (2007-07-10)
Michael A Letavic, Emily M Stocking, Ann J Barbier, Pascal Bonaventure, Jamin D Boggs, Brian Lord, Kirsten L Miller, Sandy J Wilson, Nicholas I Carruthers
ABSTRACT
The design, synthesis, and in vitro activity of a series of novel 5-ethynyl-2-aryloxybenzylamine-based histamine H(3) ligands that are also serotonin reuptake transporters is described.