MilliporeSigma
  • The synthesis and biological evaluation of a novel series of antimicrobials of the oxazolidinone class.
The synthesis and biological evaluation of a novel series of antimicrobials of the oxazolidinone class.
Bioorganic & medicinal chemistry letters (2002-07-20)
Richard J Sciotti, Marina Pliushchev, Paul E Wiedeman, Darlene Balli, Robert Flamm, Angela M Nilius, Kennan Marsh, DeAnne Stolarik, Robert Jolly, Roger Ulrich, Stevan W Djuric
ABSTRACT
A novel series of antimicrobials of the oxazolidinone class is disclosed. These compounds are characterized relative to previously described analogues by a 'halostilbene-derived' pharmacophore and demonstrate enhanced antimicrobial activity against key Gram-positive pathogens when compared to Linezolid.