MilliporeSigma
  • A protein-kinase, IFN-inducible double-stranded RNA dependent inhibitor and repressor of p58 (PRKRIR) enhances type I IFN-mediated antiviral response through the stability control of RIG-I protein.
A protein-kinase, IFN-inducible double-stranded RNA dependent inhibitor and repressor of p58 (PRKRIR) enhances type I IFN-mediated antiviral response through the stability control of RIG-I protein.
Biochemical and biophysical research communications (2011-09-14)
Hesung Now, Joo-Yeon Yoo
ABSTRACT
The cellular RIG-I-like receptor (RLR) senses pathogenic RNA molecular patterns and transmits signals for type I interferon (IFN) production. It acts as a center for antiviral responses, and large numbers of RIG-I (retinoic acid inducible gene-I) interacting proteins are identified as signaling regulators. In the present study, we report PRKRIR, a negative regulator of PKR inhibitor, as a novel RIG-I interacting protein. In HEK293FT cells, PRKRIR synergistically enhances type I IFN production mediated by a signal activated- or constitutively active form of RIG-I. The C-terminal domain of the PRKRIR was required for physical interaction and the signal augmentation. The PRKRIR blocks poly-ubiquitination and protein degradation of RIG-I, thereby increasing cellular levels of RIG-I proteins. Furthermore, overexpression of PRKRIR, along with a signal activated- or constitutively active form of RIG-I, efficiently inhibits virus replication in the infected host. In conclusion, PRKRIR provides a novel positive regulator controlling the RIG-I-IFN production system through protein stability control.