MilliporeSigma
  • Cloning of another human serotonin receptor (5-HT1F): a fifth 5-HT1 receptor subtype coupled to the inhibition of adenylate cyclase.
Cloning of another human serotonin receptor (5-HT1F): a fifth 5-HT1 receptor subtype coupled to the inhibition of adenylate cyclase.
Proceedings of the National Academy of Sciences of the United States of America (1993-01-15)
N Adham, H T Kao, L E Schecter, J Bard, M Olsen, D Urquhart, M Durkin, P R Hartig, R L Weinshank, T A Branchek
ABSTRACT
An intronless gene encoding an additional human serotonin (5-HT) 5-HT1-like receptor subtype was isolated from a human genomic library with probes obtained from degenerate PCR primers used to amplify 5-HT-receptor-specific sequences. The highest degree of homology was found with the 5-HT1E subtype (70%) and the 5-HT1D alpha (63%) and 5-HT1D beta (60%) receptors. RNA for this gene was detected in the human brain but was not detected in kidney, liver, spleen, heart, pancreas, and testes. High-affinity (Kd = 9.2 nM) 3H-labeled 5-HT binding was detected. Competition studies revealed the following rank order of potencies for serotonergic ligands: 5-HT > sumatriptan > 5-carboxyamidotryptamine > 8-hydroxy-2(di-1-propylamino)tetralin > spiperone. 5-HT produced a dose-dependent inhibition of forskolin-stimulated cAMP accumulation (EC50 = 7.9 nM) in transfected cells. These properties distinguish this receptor from any previously characterized and establish a fifth 5-HT1-like receptor subtype (5-HT1F) coupled to the inhibition of adenylate cyclase.