MilliporeSigma
  • Identification of a novel cell cycle regulated gene, HURP, overexpressed in human hepatocellular carcinoma.
Identification of a novel cell cycle regulated gene, HURP, overexpressed in human hepatocellular carcinoma.
Oncogene (2003-01-16)
Ann-Ping Tsou, Chu-Wen Yang, Chi-Ying F Huang, Ricky Chang-Tze Yu, Yuan-Chii G Lee, Cha-Wei Chang, Bo-Rue Chen, Yu-Fang Chung, Ming-Ji Fann, Chin-Wen Chi, Jen-Hwey Chiu, Chen-Kung Chou
ABSTRACT
An analytic strategy was followed to identify putative regulatory genes during the development of human hepatocellular carcinoma (HCC). This strategy employed a bioinformatics analysis that used a database search to identify genes, which are differentially expressed in human HCC and are also under cell cycle regulation. A novel cell cycle regulated gene (HURP) that is overexpressed in HCC was identified. Full-length cDNAs encoding the human and mouse HURP genes were isolated. They share 72 and 61% identity at the nucleotide level and amino-acid level, respectively. Endogenous levels of HURP mRNA were found to be tightly regulated during cell cycle progression as illustrated by its elevated expression in the G(2)/M phase of synchronized HeLa cells and in regenerating mouse liver after partial hepatectomy. Immunofluorescence studies revealed that hepatoma up-regulated protein (HURP) localizes to the spindle poles during mitosis. Overexpression of HURP in 293T cells resulted in an enhanced cell growth at low serum levels and at polyhema-based, anchorage-independent growth assay. Taken together, these results strongly suggest that HURP is a potential novel cell cycle regulator that may play a role in the carcinogenesis of human cancer cells.