MilliporeSigma
  • Histone deacetylase 5 is not a p53 target gene, but its overexpression inhibits tumor cell growth and induces apoptosis.
Histone deacetylase 5 is not a p53 target gene, but its overexpression inhibits tumor cell growth and induces apoptosis.
Cancer research (2002-05-23)
Yuanhui Huang, Mingjia Tan, Mark Gosink, Kevin K W Wang, Yi Sun
ABSTRACT
p53 tumor suppressor is activated by phosphorylation and acetylation on DNA damage. One of unknown p53 early transcripts was identified to be histone deacetylase-5 (HDAC5). We tested a hypothesis that HDAC5 is a p53 down-stream target gene that on induction by p53 inactivates p53 by removal of acetyl group in p53 molecule, thus functioning as an auto-regulatory negative feedback loop in analogue to p53-murine double minute 2 interaction. Six p53 binding consensus sites were identified in the promoter of HDAC5. p53 binds to one of the sites weakly. However, luciferase constructs driven by the HDAC5 promoter containing three to six potential binding sites were not activated by p53, nor was the expression of HDAC5 mRNA induced by p53-activating agents. Furthermore, HDAC5 does not bind to p53 nor reduces etoposide-induced p53 acetylation. Thus, HDAC5 is not a p53 target gene and may act in a p53-independent manner. We next studied the effect of HDAC5 on tumor cell growth and apoptosis. Transfection of HDAC5 inhibited growth of multiple tumor cell lines including U2OS osteogenic sarcoma cells, SY5Y neuroblastoma cells, and MCF breast carcinoma cells. The growth suppression seen in HDAC5-overexpressing cells appears to be attributable partly to a reduced growth rate as revealed by cell growth assay using 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay and mainly to spontaneous apoptosis as shown by DNA fragmentation ELISA and morphological appearance. Mechanistically, repression of three cell proliferation genes in mitogen-activated protein kinase pathway and induction of seven apoptosis-related genes were identified by microarray profiling in HDAC5-overexpressed cells. Among induced genes, four (TNFR1, TNFSF7, caspase-8, and DAPK1) were associated with the tumor necrosis factor ligand-receptor death pathway. Induction of TNFR1, TNFSF7, and caspase-8 were confirmed by Northern and Western analyses. Thus, activation of tumor necrosis factor death receptor pathway appears to be associated with HDAC5-induced spontaneous apoptosis.