MilliporeSigma
  • Synthesis and spectroscopic characterisation of BODIPY based fluorescent off-on indicators with low affinity for calcium.
Synthesis and spectroscopic characterisation of BODIPY based fluorescent off-on indicators with low affinity for calcium.
Organic & biomolecular chemistry (2005-07-21)
Nikola Basarić, Mukulesh Baruah, Wenwu Qin, Bert Metten, Mario Smet, Wim Dehaen, Noël Boens
ABSTRACT
Two fluorescent off-on Ca2+ indicators based on APTRA (o-aminophenol-N,N,O-triacetic acid) as low-affinity ligand for Ca2+ and BODIPY(4,4-difluoro-4-bora-3a,4a-diaza-s-indacene) as a fluorophore were synthesized. The new BODIPY-APTRA compounds absorb in the visible spectrum, with absorption maxima from 505 nm to 570 nm, and have fluorescence spectra that span the visible spectrum, with emission maxima ranging from 525 nm to 625 nm dependent on the substituents at the alpha-positions to the nitrogen atoms. The indicators show a large increase of the fluorescence quantum yield upon increasing Ca2+ concentration. The ground-state dissociation constants Kd estimated at 20 degrees C in 100 mM KCl aqueous buffered solution, pH 7.20, for the two complexes with Ca2+ were found to be around 100 microM.
Sigma-Aldrich
N-Boc-pyrrole, 98%