MilliporeSigma
  • Down-regulation of tumor suppressor gene FEZ1/LZTS1 in breast carcinoma involves promoter methylation and associates with metastasis.
Down-regulation of tumor suppressor gene FEZ1/LZTS1 in breast carcinoma involves promoter methylation and associates with metastasis.
Breast cancer research and treatment (2008-08-08)
Ling Chen, Zhengmao Zhu, Xiaodong Sun, Xue-Yuan Dong, Jia Wei, Feng Gu, Yu-Lan Sun, Jun Zhou, Jin-Tang Dong, Li Fu
ABSTRACT
FEZ1/LZTS1 is a tumor suppressor gene located in chromosomal band 8p22, and methylation has been identified as a mechanism for its loss of function in tumors. Chromosomal deletion at 8p22 is also frequent in breast cancer. We therefore examined whether LZTS1 plays a role in breast cancer. We analyzed expression of LZTS1 at both the RNA and protein levels, and promoter methylation in a number of primary tumors and cell lines from breast cancer. We also examined the association between LZTS1 expression and different clinicopathological parameters of breast cancer. We found that the expression of LZTS1 mRNA was reduced in 25 of 50 (50%) primary tumors and 29 of 30 (97%) breast cancer cell lines. Immunohistochemical staining showed that LZTS1 protein was absent or down-regulated in 72 (72%) of 100 primary breast carcinomas. Reduced expression of LZTS1 at either the RNA or protein level was significantly correlated with lymph node metastases (P < 0.05). DNA methylation analysis revealed that the LZTS1 gene was frequently methylated in both cell lines and primary tumors from breast cancer, and the extent of DNA methylation was correlated with reduced expression of the gene. These findings suggest that LZTS1 plays a role in the development and progression of breast cancer at least through promoter methylation-mediated transcriptional downregulation.
Sigma-Aldrich
Anti-LZTS1 antibody produced in rabbit, IgG fraction of antiserum