MilliporeSigma
Theory analysis of mass spectra of long-chain isocyanates.
Rapid communications in mass spectrometry : RCM (2008-06-05)
Dongliang Liu, Ce Hao, Hua Zhang, Weihong Qiao, Zongshi Li, Guanghui Yu, Kelu Yan, Yuliang Guo, Lvbo Cheng
ABSTRACT
Electron impact mass spectra of four long-chain isocyanates, lauryl isocyanate, tetradecyl isocyanate, hexadecyl isocyanate and octadecyl isocyanate, were obtained with a GCT high-resolution time-of-flight mass spectrometer. The four isocyanates studied gave a common base peak of m/z 99, which suggested the formation of a stable six-membered ring structure to decentralize the positive charge. Quantum-mechanical energy calculation justified that the six-membered ring base peak had the lowest energy. The positive charge assigned during the fragmentation of the radical cation, and the relative intensity of the fragment ion peaks, were explained by quantum-mechanical calculations as well.
Sigma-Aldrich
Hexadecyl isocyanate, 97%