MilliporeSigma
Chemoselective hydrosilylation of hydroxyketones.
Tetrahedron (2013-09-03)
Marta L Lage, Scott J Bader, Kanicha Sa-Ei, John Montgomery
ABSTRACT
A chemoselective method for the hydrosilylation of ketones has been developed, using the combination of triphenylsilane and a catalyst prepared from Ni(COD)