MilliporeSigma
  • Disassembly of mitotic checkpoint complexes by the joint action of the AAA-ATPase TRIP13 and p31(comet).
Disassembly of mitotic checkpoint complexes by the joint action of the AAA-ATPase TRIP13 and p31(comet).
Proceedings of the National Academy of Sciences of the United States of America (2014-08-06)
Esther Eytan, Kexi Wang, Shirly Miniowitz-Shemtov, Danielle Sitry-Shevah, Sharon Kaisari, Tim J Yen, Song-Tao Liu, Avram Hershko
ABSTRACT
The mitotic (or spindle assembly) checkpoint system delays anaphase until all chromosomes are correctly attached to the mitotic spindle. When the checkpoint is active, a Mitotic Checkpoint Complex (MCC) assembles and inhibits the ubiquitin ligase Anaphase-Promoting Complex/Cyclosome (APC/C). MCC is composed of the checkpoint proteins Mad2, BubR1, and Bub3 associated with the APC/C activator Cdc20. When the checkpoint signal is turned off, MCC is disassembled and the checkpoint is inactivated. The mechanisms of the disassembly of MCC are not sufficiently understood. We have previously observed that ATP hydrolysis is required for the action of the Mad2-binding protein p31(comet) to disassemble MCC. We now show that HeLa cell extracts contain a factor that promotes ATP- and p31(comet)-dependent disassembly of a Cdc20-Mad2 subcomplex and identify it as Thyroid Receptor Interacting Protein 13 (TRIP13), an AAA-ATPase known to interact with p31(comet). The joint action of TRIP13 and p31(comet) also promotes the release of Mad2 from MCC, participates in the complete disassembly of MCC and abrogates checkpoint inhibition of APC/C. We propose that TRIP13 plays centrally important roles in the sequence of events leading to MCC disassembly and checkpoint inactivation.