MilliporeSigma
  • Mospd1, a new player in mesenchymal versus epidermal cell differentiation.
Mospd1, a new player in mesenchymal versus epidermal cell differentiation.
Journal of cellular physiology (2011-07-28)
R Thaler, M Rumpler, S Spitzer, K Klaushofer, F Varga
ABSTRACT
Mospd1 codes for a small protein with unknown physiological function, which is part of a family of genes, including Mospd2 and Mospd3, defined by the presence of the major sperm protein domain and two transmembrane domains. This work characterizes the Mospd1 gene, the intracellular location of the protein and its expression in different mouse tissues and mesenchymal cell lines during differentiation. The role of Mospd1 in mesenchymal cellular differentiation was studied by siRNA knockdown experiments in mouse osteoblastic MC3T3-E1 cells. Transfection experiments of the targeted cDNA show MOSPD1 located in the endoplasmatic reticulum and in the Golgi apparatus. Removal of the last exon of the gene resulted in localization of the protein in the nucleus, which was attributed to a nuclear export sequence in the N-terminal part. In mouse tissues the gene was generally strongly expressed while mesenchymal tissues showed the highest expression. In mesenchymal cell lines Mospd1 mRNA was higher expressed in cells with advanced differentiation status. In osteoblastic, myoblastic, and adipocytic cell lines Mospd1 was up-regulated during differentiation. Genome-wide gene expression analysis after knockdown of Mospd1 by siRNA in MC3T3-E1 cells revealed a shift in the gene expression pattern from mesenchymal to epithelial genes featuring up-regulation of the epithelial cadherin Cdh1 and down-regulation of its inhibitors Snail1 and 2 and the mesenchymal cadherin Cdh11, suggesting a mesenchymal to epithelial transition. From these data we conclude that Mospd1 plays a pivotal role in the developmental regulation at the switch between mesenchymal and epithelial cells.