MilliporeSigma
  • ZnO-CuO core-branch nanocatalysts for ultrasound-assisted azide-alkyne cycloaddition reactions.
ZnO-CuO core-branch nanocatalysts for ultrasound-assisted azide-alkyne cycloaddition reactions.
Chemical communications (Cambridge, England) (2012-05-05)
Ji Chan Park, A Young Kim, Jae Young Kim, Sungkyun Park, Kang Hyun Park, Hyunjoon Song
ABSTRACT
ZnO-CuO core-branch hybrid nanoparticles, synthesized by copper oxide growth and controlled oxidation on ZnO nanospheres, exhibited remarkable enhancement of catalytic activity and stability for ultrasound-assisted [3+2] azide-alkyne cycloaddition reactions, due to their high surface area and active facets of the CuO branches.
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)