MilliporeSigma
  • Actinin-associated LIM protein: identification of a domain interaction between PDZ and spectrin-like repeat motifs.
Actinin-associated LIM protein: identification of a domain interaction between PDZ and spectrin-like repeat motifs.
The Journal of cell biology (1997-10-23)
H Xia, S T Winokur, W L Kuo, M R Altherr, D S Bredt
ABSTRACT
PDZ motifs are protein-protein interaction domains that often bind to COOH-terminal peptide sequences. The two PDZ proteins characterized in skeletal muscle, syntrophin and neuronal nitric oxide synthase, occur in the dystrophin complex, suggesting a role for PDZ proteins in muscular dystrophy. Here, we identify actinin-associated LIM protein (ALP), a novel protein in skeletal muscle that contains an NH2-terminal PDZ domain and a COOH-terminal LIM motif. ALP is expressed at high levels only in differentiated skeletal muscle, while an alternatively spliced form occurs at low levels in the heart. ALP is not a component of the dystrophin complex, but occurs in association with alpha-actinin-2 at the Z lines of myofibers. Biochemical and yeast two-hybrid analyses demonstrate that the PDZ domain of ALP binds to the spectrin-like motifs of alpha-actinin-2, defining a new mode for PDZ domain interactions. Fine genetic mapping studies demonstrate that ALP occurs on chromosome 4q35, near the heterochromatic locus that is mutated in fascioscapulohumeral muscular dystrophy.