MilliporeSigma
  • ZNF75: isolation of a cDNA clone of the KRAB zinc finger gene subfamily mapped in YACs 1 Mb telomeric of HPRT.
ZNF75: isolation of a cDNA clone of the KRAB zinc finger gene subfamily mapped in YACs 1 Mb telomeric of HPRT.
Genomics (1993-11-01)
A Villa, I Zucchi, G Pilia, D Strina, L Susani, F Morali, C Patrosso, A Frattini, F Lucchini, M Repetto
ABSTRACT
We have previously mapped a zinc finger genomic motif (ZNF75) to the Xq26 cytogenetic band by using a hybrid panel. Here, we report the isolation of the transcribed counterpart in a cDNA clone and its further localization. The cDNA clone, from a lung fibroblast library, is assembled from three exons, including a 289 amino acid (AA) long open reading frame containing a recently described motif, the Kruppel-associated box, 42 AA long, in exon 2. By comparison with other reported members of the subfamily, the exon-intron boundaries also appear to be very well conserved. Further analysis allowed us to map this gene 1 Mb downstream from the HPRT gene in the published YAC contig that extends across Xq26. Two other motifs, 87 and 78% homologous to ZNF75 at the amino acid level, were identified by PCR on total human DNA, but map outside Xq24-qter.