MilliporeSigma
  • Characterization of a cluster of sulfatase genes on Xp22.3 suggests gene duplications in an ancestral pseudoautosomal region.
Characterization of a cluster of sulfatase genes on Xp22.3 suggests gene duplications in an ancestral pseudoautosomal region.
Human molecular genetics (1996-04-01)
G Meroni, B Franco, N Archidiacono, S Messali, G Andolfi, M Rocchi, A Ballabio
ABSTRACT
An obligatory crossing-over event between the X and Y chromosomes in mammals occurs at each male meiosis within the 2.6 Mb of DNA defining the pseudoautosomal region (PAR). Genes located within or near the human PAR have homologous copies on the X and Y chromosomes, escape X inactivation and appear to be highly divergent throughout evolution. We have characterized the genomic structure of two genes from a recently identified cluster of sulfatase genes (ARSD and ARSE) located in the Xp22.3 region, and of their homologs on the Y chromosome. Our results indicate that the ARSD and ARSE genes from within this cluster have a conserved genomic organization, shared also by another Xp22.3 gene, STS, but completely different from that of all the other sulfatase genes. Sequence analysis of the Y-linked homologs indicate that they represent truncated pseudogenes. Sequence identity values between the X and Y copies of each gene is on average 91%, significantly higher than the values obtained by comparing different members of the family. FISH mapping experiments performed in several primate species revealed an identical localization of the X-linked copies to that in man, but different localizations of the Y homologs. Together, our data indicate that the cluster of sulfatase genes on human Xp22.3 was created through duplication events which probably occurred in an ancestral PAR, and support the view that the PAR has undergone multiple changes during recent mammalian evolution.