MilliporeSigma
  • MCM7 serves as a prognostic marker in diffuse-type gastric adenocarcinoma and siRNA-mediated knockdown suppresses its oncogenic function.
MCM7 serves as a prognostic marker in diffuse-type gastric adenocarcinoma and siRNA-mediated knockdown suppresses its oncogenic function.
Oncology reports (2014-03-22)
Wei Kang, Joanna H M Tong, Anthony W H Chan, Alfred S L Cheng, Jun Yu, Kafai To
ABSTRACT
MCM7 (mini-chromosome maintenance protein 7) is essential for the initiation of genomic replication as a component of the pre-replication complex. The present study aimed to analyze its expression, clinical significance and biological functions in gastric adenocarcinoma (GAC). The MCM7 protein was upregulated in all 9 GAC cell lines. In 6 paired primary GACs, MCM7 was upregulated in tumor compared with the corresponding non-tumorous gastric tissues. In normal gastric epithelium tissue, MCM7 was strictly expressed in the proliferative compartment. MCM7 knockdown by siRNA in gastric cancer cell line AGS and NCI-N87 significantly suppressed cell proliferation, inhibited monolayer colony formation, reduced cell invasion and induced late apoptosis. Its nuclear expression correlated with advanced age and poorer disease specific survival in diffuse-type GACs. All the findings supported that MCM7 might play an oncogenic role in gastric tumorigenesis. It serves as a potential prognostic marker and therapeutic target in diffuse-type GACs.