MilliporeSigma
  • Activation of the cAMP cascade in human fibroblast cultures rescues the activity of oxidatively damaged complex I.
Activation of the cAMP cascade in human fibroblast cultures rescues the activity of oxidatively damaged complex I.
Free radical biology & medicine (2011-12-27)
Domenico De Rasmo, Anna Signorile, Maria Larizza, Consiglia Pacelli, Tiziana Cocco, Sergio Papa
ABSTRACT
A study of the relationship between cAMP/PKA-dependent phosphorylation and oxidative damage of subunits of complex I of the mitochondrial respiratory chain is presented. It is shown that, in fibroblast cultures, PKA-mediated phosphorylation of the NDUFS4 subunit of complex I rescues the activity of the oxidatively damaged complex. Evidence is presented showing that this effect is mediated by phosphorylation-dependent exchange of carbonylated NDUFS4 subunit in the assembled complex with the de novo synthesized subunit. These results indicate a potential use for β-adrenoceptor agonists in preventing/reversing the detrimental effects of oxidative stress in the mitochondrial respiratory system.