MilliporeSigma
  • 2,3 Dimercapto-1-propanol inhibits HIV-1 tat activity, viral production, and infectivity in vitro.
2,3 Dimercapto-1-propanol inhibits HIV-1 tat activity, viral production, and infectivity in vitro.
AIDS research and human retroviruses (1990-07-01)
S Kubota, M A el-Farrash, M Maki, S Harada, M Hatanaka
ABSTRACT
We have examined the effect of 2,3 dimercapto-1-propanol (DMP), which is known as an anti-heavy metal-poisoning drug, against human immunodeficiency virus type 1 (HIV-1). We demonstrate that DMP inhibited transactivation directed by tat protein, which is a metal containing transcriptional transactivating factor and also interfered with viral production. Furthermore, treatment and pretreatment of cells with DMP strongly reduced their sensitivity for HIV-1 infection through unknown mechanisms. These results indicate that DMP reveals pleuripotent effects on HIV-1 infection and production in vitro and thus may provide an exploitable hypothesis for designing new drugs against AIDS.
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)