MilliporeSigma
  • Identification and characterization of a novel cancer/testis antigen gene CAGE.
Identification and characterization of a novel cancer/testis antigen gene CAGE.
Biochemical and biophysical research communications (2002-04-02)
Bomsoo Cho, Yoon Lim, Dae-Yeon Lee, Sae-Young Park, Hosoon Lee, Woo Ho Kim, Hankwang Yang, Yung-Jue Bang, Doo-Il Jeoung
ABSTRACT
We applied serological analysis of cDNA expression library technique to identify cancer-associated genes. We screened cDNA expression libraries of human testis and gastric cancer cell lines with sera of patients with gastric cancers. We identified a gene whose expression is testis-specific among normal tissues. We cloned and characterized this novel gene. It contains D-E-A-D box domain and encodes a putative protein of 630 amino acids with possible helicase activity. It showed wide expression in various cancer tissues and cancer cell lines. The corresponding gene was named cancer-associated gene (CAGE). PCR of human x hamster Radiation Hybrids showed localization of CAGE on the human chromosome Xp22. Transient transfection of CAGE showed predominantly nuclear localization. Both Western blot and plaque assay indicated seroreactivity of CAGE protein. We found that demethylation played a role in the activation of CAGE in some cancer cell lines that do not express it. Cell synchronization experiments showed that the expression of CAGE was related with cell cycle. This suggests that CAGE might play a role in cellular proliferation. Because CAGE is expressed in a variety of cancers but not in normal tissues except testis, this gene can be a target of antitumor immunotherapy.