MilliporeSigma
  • Synthesis and evaluation of the antiparasitic activity of bis-(arylmethylidene) cycloalkanones.
Synthesis and evaluation of the antiparasitic activity of bis-(arylmethylidene) cycloalkanones.
European journal of medicinal chemistry (2013-12-11)
Saulo F P Braga, Érika V P Alves, Rafaela S Ferreira, Jordana R B Fradico, Paula S Lage, Mariana C Duarte, Tatiana G Ribeiro, Policarpo A S Júnior, Alvaro J Romanha, Maiko L Tonini, Mário Steindel, Eduardo F Coelho, Renata B de Oliveira
ABSTRACT
A series of bis-(arylmethylidene)-cycloalkanones was synthesized by cross-aldol condensation. The activity of the compounds was evaluated against amastigotes forms of Trypanosoma cruzi and promastigotes forms of Leishmania amazonensis. The cytotoxicity of the active compounds on uninfected fibroblasts or macrophages was established in vitro to evaluate the selectivity of their antiparasitic effects. Six compounds displayed trypanocidal activity against amastigotes intracellular forms of T. cruzi with IC₅₀ values ranging from 7.0 to 249 μM. Besides these six compounds, eight other molecules exhibited significant leishmanicidal activity (IC₅₀ values ranging from 0.6 to 110.4 μM). Two compounds can be considered as promising antiparasitic lead molecules because they showed IC₅₀ values in the low-micromolar range (≤1.2 μM) with an adequate SI (≥19.9). To understand the mechanism of action of these compounds, two possible molecular targets were investigated: trypanothione reductase (TR) and cruzain.
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)
Supelco
Cyclohexanone, analytical standard
Sigma-Aldrich
Cyclopentanone, ReagentPlus®, ≥99%