MilliporeSigma
  • Characterization of a 95 kDa high affinity human high density lipoprotein-binding protein.
Characterization of a 95 kDa high affinity human high density lipoprotein-binding protein.
Biochemistry (2001-04-04)
A V Bocharov, T G Vishnyakova, I N Baranova, A P Patterson, T L Eggerman
ABSTRACT
A new human 95 kDa high density lipoprotein (HDL)-binding protein (HBP) corresponding to a high affinity HDL-binding site with K(d) = 1.67 microg/mL and a capacity of 13.4 ng/mg was identified in human fetal hepatocytes. The HDL binding with the 95 kDa HBP plateaus at 2.5-5 microg/mL under reducing and nonreducing conditions. The association of HDL(3) with the 95 kDa HBP plateaued in 15-30 min while dissociation was complete in 30 min. HDL(3), apoA-I, and apoA-II were recognized by the 95 kDa HBP while low density lipoproteins (LDL) and tetranitromethane-modified HDL were not. The 95 kDa HBP predominantly resides on the surface of cells since trypsin treatment of HepG2 cells eliminated nearly 70% of HDL binding. All studied human cells and cell lines (HepG2, Caco-2, HeLa, fibroblasts, SKOV-3, PA-I) demonstrated the presence of the 95 kDa protein. Both RT-PCR and Western blotting for HB-2/ALCAM were negative in human fetal hepatocytes while Gp96/GRP94 was clearly differentiated from the 95 kDa HBP by two-dimensional electrophoretic mobility. Moreover, deglycosylation of HepG2 membrane preparations did not affect either HDL binding to the 95 kDa HBP or its size, while in contrast it affected the molecular weights of HB-2/ALCAM and SR-BI/CLA-1. We conclude that the 95 kDa HBP is a new HDL receptor candidate widely expressed in human cells and cell lines.