MilliporeSigma
  • Platinum-catalyzed tandem indole annulation/arylation for the synthesis of diindolylmethanes and indolo[3,2-b]carbazoles.
Platinum-catalyzed tandem indole annulation/arylation for the synthesis of diindolylmethanes and indolo[3,2-b]carbazoles.
Organic letters (2013-08-06)
Dongxu Shu, Gabrielle N Winston-McPherson, Wangze Song, Weiping Tang
ABSTRACT
Various diindolylmethanes were prepared from propargylic ethers and substituted indoles via a platinum-catalyzed tandem indole annulation/arylation cascade. The resulting diindolylmethanes could be converted to natural product malassezin by formylation or indolo[3,2-b]carbazoles by cyclization.