MilliporeSigma
Organization of the human myoglobin gene.
The EMBO journal (1984-02-01)
P Weller, A J Jeffreys, V Wilson, A Blanchetot
ABSTRACT
Cross-hybridization of the grey seal myoglobin gene to human DNA detected a single human myoglobin gene plus an extensive family of sequences apparently related to the central exon of this gene. The functional human gene is 10.4 kb long and has a haemoglobin-like three exon/two intron structure with long non-coding regions similar to its seal homologue. At least 300 bp of 5'-flanking region are closely homologous between the two genes, with the exception of a divergent purine-rich region 68-114 bp upstream of the cap site. A diverged tandem repetitive sequence based on (GGAT)165 is located 1100-1750 bp upstream from the gene; internal homology units within this sequence suggest sequence homogenization by gene microconversions. A second 33-bp tandem repeat element in the first intron is flanked by a 9-bp direct repeat, shares homology with other tandem repetitive elements in the human genome and may represent a novel form of transposable element.