MilliporeSigma
  • Acylcarnitine removal in a patient with acyl-CoA beta-oxidation deficiency disorder: effect of L-carnitine therapy and starvation.
Acylcarnitine removal in a patient with acyl-CoA beta-oxidation deficiency disorder: effect of L-carnitine therapy and starvation.
Clinica chimica acta; international journal of clinical chemistry (1996-08-30)
M Fontaine, G Briand, L Vallée, G Ricart, P Degand, P Divry, C Vianey-Saban, J Vamecq
ABSTRACT
Carnitine levels and acylcarnitine profiles in a patient with mild multiple acyl-CoA dehydrogenase deficient beta-oxidation were compared with control results. Whereas blood and urine total carnitine levels were moderately decreased, blood esterified carnitine levels in the patient were about 2-fold higher than in controls. Urinary acylcarnitine profiles presented with a larger variety of carnitine esters than in controls and included propionylcarnitine, butyrylcarnitine, 2-methylbutyrylcarnitine, hexanoylcarnitine and octanolycarnitine. Total carnitine levels in body fluids were similarly affected by chronic oral L-carnitine administration in patient and controls. By contrast, esterified carnitine level increase was 2-fold more important in controls than in patient. Whereas no qualitative changes in urinary acylcarnitine profiles were induced by L-carnitine therapy in controls, several alterations of these profiles were observed in the patient. The effect of starvation on metabolites was also studied, especially beta-oxidation rates assessed by free fatty acids to 3-hydroxybutyric acid ratios in blood from the patient in the untreated and L-carnitine treated states. In the L-carnitine-supplemented patient, the effect of starvation on the time course of carnitine levels and acylcarnitine profiles could also be documented. The ability of chronic oral L-carnitine administration to remove relatively less important amounts of acylcarnitines in the patient than in controls is further discussed, as well as qualitative alterations of acylcarnitine profiles induced by this therapy in the pathological condition.
Sigma-Aldrich
2-Methylbutyryl-L-carnitine, ≥97.0% (HPLC)