MilliporeSigma
  • IQCJ-SCHIP1, a novel fusion transcript encoding a calmodulin-binding IQ motif protein.
IQCJ-SCHIP1, a novel fusion transcript encoding a calmodulin-binding IQ motif protein.
Biochemical and biophysical research communications (2006-10-19)
Dorota A Kwaśnicka-Crawford, Andrew R Carson, Stephen W Scherer
ABSTRACT
The existence of transcripts that span two adjacent, independent genes is considered rare in the human genome. This study characterizes a novel human fusion gene named IQCJ-SCHIP1. IQCJ-SCHIP1 is the longest isoform of a complex transcriptional unit that bridges two separate genes that encode distinct proteins, IQCJ, a novel IQ motif containing protein and SCHIP1, a schwannomin interacting protein that has been previously shown to interact with the Neurofibromatosis type 2 (NF2) protein. IQCJ-SCHIP1 is located on the chromosome 3q25 and comprises a 1692-bp transcript encompassing 11 exons spanning 828kb of the genomic DNA. We show that IQCJ-SCHIP1 mRNA is highly expressed in the brain. Protein encoded by the IQCJ-SCHIP1 gene was localized to cytoplasm and actin-rich regions and in differentiated PC12 cells was also seen in neurite extensions.