MilliporeSigma
  • Broad and direct interaction between TLR and Siglec families of pattern recognition receptors and its regulation by Neu1.
Broad and direct interaction between TLR and Siglec families of pattern recognition receptors and its regulation by Neu1.
eLife (2014-09-05)
Guo-Yun Chen, Nicholas K Brown, Wei Wu, Zahra Khedri, Hai Yu, Xi Chen, Diantha van de Vlekkert, Alessandra D'Azzo, Pan Zheng, Yang Liu
ABSTRACT
Both pathogen- and tissue damage-associated molecular patterns induce inflammation through toll-like receptors (TLRs), while sialic acid-binding immunoglobulin superfamily lectin receptors (Siglecs) provide negative regulation. Here we report extensive and direct interactions between these pattern recognition receptors. The promiscuous TLR binders were human SIGLEC-5/9 and mouse Siglec-3/E/F. Mouse Siglec-G did not show appreciable binding to any TLRs tested. Correspondingly, Siglece deletion enhanced dendritic cell responses to all microbial TLR ligands tested, while Siglecg deletion did not affect the responses to these ligands. TLR4 activation triggers Neu1 translocation to cell surface to disrupt TLR4:Siglec-E interaction. Conversely, sialidase inhibitor Neu5Gc2en prevented TLR4 ligand-induced disruption of TLR4:Siglec E/F interactions. Absence of Neu1 in hematopoietic cells or systematic treatment with sialidase inhibitor Neu5Gc2en protected mice against endotoxemia. Our data raised an intriguing possibility of a broad repression of TLR function by Siglecs and a sialidase-mediated de-repression that allows positive feedback of TLR activation during infection.
Sigma-Aldrich
2′-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid sodium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Neu1