MilliporeSigma
  • Induction of body weight loss through RNAi-knockdown of APOBEC1 gene expression in transgenic rabbits.
Induction of body weight loss through RNAi-knockdown of APOBEC1 gene expression in transgenic rabbits.
PloS one (2014-09-13)
Geneviève Jolivet, Sandrine Braud, Bruno DaSilva, Bruno Passet, Erwana Harscoët, Céline Viglietta, Thomas Gautier, Laurent Lagrost, Nathalie Daniel-Carlier, Louis-Marie Houdebine, Itzik Harosh
ABSTRACT
In the search of new strategies to fight against obesity, we targeted a gene pathway involved in energy uptake. We have thus investigated the APOB mRNA editing protein (APOBEC1) gene pathway that is involved in fat absorption in the intestine. The APOB gene encodes two proteins, APOB100 and APOB48, via the editing of a single nucleotide in the APOB mRNA by the APOBEC1 enzyme. The APOB48 protein is mandatory for the synthesis of chylomicrons by intestinal cells to transport dietary lipids and cholesterol. We produced transgenic rabbits expressing permanently and ubiquitously a small hairpin RNA targeting the rabbit APOBEC1 mRNA. These rabbits exhibited a moderately but significantly reduced level of APOBEC1 gene expression in the intestine, a reduced level of editing of the APOB mRNA, a reduced level of synthesis of chylomicrons after a food challenge, a reduced total mass of body lipids and finally presented a sustained lean phenotype without any obvious physiological disorder. Interestingly, no compensatory mechanism opposed to the phenotype. These lean transgenic rabbits were crossed with transgenic rabbits expressing in the intestine the human APOBEC1 gene. Double transgenic animals did not present any lean phenotype, thus proving that the intestinal expression of the human APOBEC1 transgene was able to counterbalance the reduction of the rabbit APOBEC1 gene expression. Thus, a moderate reduction of the APOBEC1 dependent editing induces a lean phenotype at least in the rabbit species. This suggests that the APOBEC1 gene might be a novel target for obesity treatment.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Anti-APOBEC1 antibody produced in rabbit, affinity isolated antibody