MilliporeSigma
  • Global analysis of neutrophil responses to Neisseria gonorrhoeae reveals a self-propagating inflammatory program.
Global analysis of neutrophil responses to Neisseria gonorrhoeae reveals a self-propagating inflammatory program.
PLoS pathogens (2014-09-05)
Anna Sintsova, Helen Sarantis, Epshita A Islam, Chun Xiang Sun, Mohsen Amin, Carlos H F Chan, Clifford P Stanners, Michael Glogauer, Scott D Gray-Owen
ABSTRACT
An overwhelming neutrophil-driven response causes both acute symptoms and the lasting sequelae that result from infection with Neisseria gonorrhoeae. Neutrophils undergo an aggressive opsonin-independent response to N. gonorrhoeae, driven by the innate decoy receptor CEACAM3. CEACAM3 is exclusively expressed by human neutrophils, and drives a potent binding, phagocytic engulfment and oxidative killing of Opa-expressing bacteria. In this study, we sought to explore the contribution of neutrophils to the pathogenic inflammatory process that typifies gonorrhea. Genome-wide microarray and biochemical profiling of gonococcal-infected neutrophils revealed that CEACAM3 engagement triggers a Syk-, PKCδ- and Tak1-dependent signaling cascade that results in the activation of an NF-κB-dependent transcriptional response, with consequent production of pro-inflammatory cytokines. Using an in vivo model of N. gonorrhoeae infection, we show that human CEACAM-expressing neutrophils have heightened migration toward the site of the infection where they may be further activated upon Opa-dependent binding. Together, this study establishes that the role of CEACAM3 is not restricted to the direct opsonin-independent killing by neutrophils, since it also drives the vigorous inflammatory response that typifies gonorrhea. By carrying the potential to mobilize increasing numbers of neutrophils, CEACAM3 thereby represents the tipping point between protective and pathogenic outcomes of N. gonorrhoeae infection.
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%