MilliporeSigma
  • Adhesin competence repressor (AdcR) from Streptococcus pyogenes controls adaptive responses to zinc limitation and contributes to virulence.
Adhesin competence repressor (AdcR) from Streptococcus pyogenes controls adaptive responses to zinc limitation and contributes to virulence.
Nucleic acids research (2014-12-17)
Misu Sanson, Nishanth Makthal, Anthony R Flores, Randall J Olsen, James M Musser, Muthiah Kumaraswami
ABSTRACT
Altering zinc bioavailability to bacterial pathogens is a key component of host innate immunity. Thus, the ability to sense and adapt to the alterations in zinc concentrations is critical for bacterial survival and pathogenesis. To understand the adaptive responses of group A Streptococcus (GAS) to zinc limitation and its regulation by AdcR, we characterized gene regulation by AdcR. AdcR regulates the expression of 70 genes involved in zinc acquisition and virulence. Zinc-bound AdcR interacts with operator sequences in the negatively regulated promoters and mediates differential regulation of target genes in response to zinc deficiency. Genes involved in zinc mobilization and conservation are derepressed during mild zinc deficiency, whereas the energy-dependent zinc importers are upregulated during severe zinc deficiency. Further, we demonstrated that transcription activation by AdcR occurs by direct binding to the promoter. However, the repression and activation by AdcR is mediated by its interactions with two distinct operator sequences. Finally, mutational analysis of the metal ligands of AdcR caused impaired DNA binding and attenuated virulence, indicating that zinc sensing by AdcR is critical for GAS pathogenesis. Together, we demonstrate that AdcR regulates GAS adaptive responses to zinc limitation and identify molecular components required for GAS survival during zinc deficiency.
Product Description
Zinc, foil, 25mm disks, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 1m coil, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 50mm disks, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 1.0mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 4mm disks, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, 50mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 5m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.01mm, 99.9%
Zinc, foil, 6mm disks, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 2m coil, thickness 1.0mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.015mm, 99.9%
Zinc, foil, 6mm disks, thickness 0.005mm, 99.9%
Zinc, foil, 2m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Zinc, foil, 4mm disks, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.0025mm, 99.9%
Zinc, foil, 8mm disks, thickness 0.02mm, 99.9%
Zinc, foil, 2m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.95+%