MilliporeSigma
  • Mutations in G protein β subunits promote transformation and kinase inhibitor resistance.
Mutations in G protein β subunits promote transformation and kinase inhibitor resistance.
Nature medicine (2014-12-09)
Akinori Yoda, Guillaume Adelmant, Jerome Tamburini, Bjoern Chapuy, Nobuaki Shindoh, Yuka Yoda, Oliver Weigert, Nadja Kopp, Shuo-Chieh Wu, Sunhee S Kim, Huiyun Liu, Trevor Tivey, Amanda L Christie, Kutlu G Elpek, Joseph Card, Kira Gritsman, Jason Gotlib, Michael W Deininger, Hideki Makishima, Shannon J Turley, Nathalie Javidi-Sharifi, Jaroslaw P Maciejewski, Siddhartha Jaiswal, Benjamin L Ebert, Scott J Rodig, Jeffrey W Tyner, Jarrod A Marto, David M Weinstock, Andrew A Lane
ABSTRACT
Activating mutations in genes encoding G protein α (Gα) subunits occur in 4-5% of all human cancers, but oncogenic alterations in Gβ subunits have not been defined. Here we demonstrate that recurrent mutations in the Gβ proteins GNB1 and GNB2 confer cytokine-independent growth and activate canonical G protein signaling. Multiple mutations in GNB1 affect the protein interface that binds Gα subunits as well as downstream effectors and disrupt Gα interactions with the Gβγ dimer. Different mutations in Gβ proteins clustered partly on the basis of lineage; for example, all 11 GNB1 K57 mutations were in myeloid neoplasms, and seven of eight GNB1 I80 mutations were in B cell neoplasms. Expression of patient-derived GNB1 variants in Cdkn2a-deficient mouse bone marrow followed by transplantation resulted in either myeloid or B cell malignancies. In vivo treatment with the dual PI3K-mTOR inhibitor BEZ235 suppressed GNB1-induced signaling and markedly increased survival. In several human tumors, mutations in the gene encoding GNB1 co-occurred with oncogenic kinase alterations, including the BCR-ABL fusion protein, the V617F substitution in JAK2 and the V600K substitution in BRAF. Coexpression of patient-derived GNB1 variants with these mutant kinases resulted in inhibitor resistance in each context. Thus, GNB1 and GNB2 alterations confer transformed and resistance phenotypes across a range of human tumors and may be targetable with inhibitors of G protein signaling.
Sigma-Aldrich
Adenosine 5′-Triphosphatase from porcine cerebral cortex, lyophilized powder, ≥0.3 units/mg protein, pH 7.8
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)