MilliporeSigma
  • Ultrasound radiation from a three-layer thermoacoustic transformation device.
Ultrasound radiation from a three-layer thermoacoustic transformation device.
Ultrasonics (2014-12-04)
Takuya Nishioka, Yu Teshima, Takashi Mano, Ken Sakai, Takaaki Asada, Mami Matsukawa, Tetsuo Ohta, Shizuko Hiryu
ABSTRACT
A thermophone is a thermoacoustic transducer, which generates sound via time-varying Joule heating of an electrically conductive layer, which leads to expansion and contraction of a small pocket of air near the surface of the film. In this work, a 10-μm-thick Ag-Pd conductive film was coupled with heat-insulating and heat-releasing layers to fabricate a three-layer thermophone for generating ultrasound. The heat-insulating layer was 47 μm thick, and was made of glass. The heat-releasing layer was 594 μm thick, and was made of 94% alumina. Because of the simple sound-generation mechanism, which does not require mechanical moving parts, the Ag-Pd conductive film on the glass substrate can produce ultrasound radiation with broadband frequency characteristics, where exiting commercial electrode materials were used. We also demonstrate that the measured directivity patterns are in good agreement with theoretical predictions, assuming a rectangular diaphragm with the same size as the metallic film.
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Silver, rod, 100mm, diameter 10.0mm, as drawn, 99.95+%
Silver, foil, not light tested, 100x100mm, thickness 0.0025mm, 99.97%
Lead, foil, 1m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.55mm, as rolled, 100%
Silver, foil, 6mm disks, thickness 0.015mm, 99.97%
Lead, foil, 10mm disks, thickness 0.008mm, 99.99+%
Silver, foil, 25mm disks, thickness 0.25mm, as rolled, 99.95+%
Lead, foil, 0.5m coil, thickness 0.05mm, as rolled, 97% (sn 1.5%, sb 1.5%)