MilliporeSigma
  • Silver nanoparticles affect on gene expression of inflammatory and neurodegenerative responses in mouse brain neural cells.
Silver nanoparticles affect on gene expression of inflammatory and neurodegenerative responses in mouse brain neural cells.
Environmental research (2014-12-03)
Chin-Lin Huang, I-Lun Hsiao, Ho-Chen Lin, Chu-Fang Wang, Yuh-Jeen Huang, Chun-Yu Chuang
ABSTRACT
Silver nanoparticles (AgNPs) have antibacterial characteristics, and currently are applied in Ag-containing products. This study found neural cells can uptake 3-5 nm AgNPs, and investigated the potential effects of AgNPs on gene expression of inflammation and neurodegenerative disorder in murine brain ALT astrocytes, microglial BV-2 cells and neuron N2a cells. After AgNPs (5, 10, 12.5 μg/ml) exposure, these neural cells had obviously increased IL-1β secretion, and induced gene expression of C-X-C motif chemokine 13 (CXCL13), macrophage receptor with collagenous structure (MARCO) and glutathione synthetase (GSS) for inflammatory response and oxidative stress neutralization. Additionally, this study found amyloid-β (Aβ) plaques for pathological feature of Alzheimer's disease (AD) deposited in neural cells after AgNPs treatment. After AgNPs exposure, the gene expression of amyloid precursor protein (APP) was induced, and otherwise, neprilysin (NEP) and low-density lipoprotein receptor (LDLR) were reduced in neural cells as well as protein level. These results suggested AgNPs could alter gene and protein expressions of Aβ deposition potentially to induce AD progress in neural cells. It's necessary to take notice of AgNPs distribution in the environment.
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Silver, foil, light tested, 50x50mm, thickness 0.02mm, 99.97%
Silver, foil, light tested, 50x50mm, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 10m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, light tested, 100x100mm, thickness 0.02mm, 99.97%
Silver, foil, 5m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.99+%
Silver, foil, not light tested, 100x100mm, thickness 0.006mm, 99.97%
Silver, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, 1m coil, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 1m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, 1m coil, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 2m coil, thickness 0.0125mm, 99.95+%
Silver, wire reel, 500m, diameter 0.05mm, annealed, 99.99%
Silver, wire reel, 1m, diameter 0.015mm, hard, 99.99%
Silver, wire reel, 10m, diameter 0.05mm, annealed, 99.99%
Silver, tube, 500mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.95+%
Silver, wire reel, 500m, diameter 0.125mm, annealed, 99.99%
Silver, wire reel, 1m, diameter 0.025mm, as drawn, 99.99%
Silver, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.997%
Silver, wire reel, 5m, diameter 0.25mm, annealed, 99.99%
Silver, wire reel, 10m, diameter 0.125mm, annealed, 99.99%
Silver, wire reel, 50m, diameter 0.05mm, annealed, 99.99%