MilliporeSigma
  • The Arabidopsis COBRA protein facilitates cellulose crystallization at the plasma membrane.
The Arabidopsis COBRA protein facilitates cellulose crystallization at the plasma membrane.
The Journal of biological chemistry (2014-10-22)
Nadav Sorek, Hagit Sorek, Aleksandra Kijac, Heidi J Szemenyei, Stefan Bauer, Kian Hématy, David E Wemmer, Chris R Somerville
ABSTRACT
Mutations in the Arabidopsis COBRA gene lead to defects in cellulose synthesis but the function of COBRA is unknown. Here we present evidence that COBRA localizes to discrete particles in the plasma membrane and is sensitive to inhibitors of cellulose synthesis, suggesting that COBRA and the cellulose synthase complex reside in close proximity on the plasma membrane. Live-cell imaging of cellulose synthesis indicated that, once initiated, cellulose synthesis appeared to proceed normally in the cobra mutant. Using isothermal calorimetry, COBRA was found to bind individual β1-4-linked glucan chains with a KD of 3.2 μm. Competition assays suggests that COBRA binds individual β1-4-linked glucan chains with higher affinity than crystalline cellulose. Solid-state nuclear magnetic resonance studies of the cell wall of the cobra mutant also indicated that, in addition to decreases in cellulose amount, the properties of the cellulose fibrils and other cell wall polymers differed from wild type by being less crystalline and having an increased number of reducing ends. We interpret the available evidence as suggesting that COBRA facilitates cellulose crystallization from the emerging β1-4-glucan chains by acting as a "polysaccharide chaperone."
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, acid washed, powder, for column chromatography
Supelco
Cellulose, powder, for column chromatography
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Supelco
Cellulose, acid washed, from spruce, for column chromatography