MilliporeSigma
  • Phase I study of cabazitaxel plus cisplatin in patients with advanced solid tumors: study to evaluate the impact of cytochrome P450 3A inhibitors (aprepitant, ketoconazole) or inducers (rifampin) on the pharmacokinetics of cabazitaxel.
Phase I study of cabazitaxel plus cisplatin in patients with advanced solid tumors: study to evaluate the impact of cytochrome P450 3A inhibitors (aprepitant, ketoconazole) or inducers (rifampin) on the pharmacokinetics of cabazitaxel.
Cancer chemotherapy and pharmacology (2014-10-14)
John Sarantopoulos, Alain C Mita, James L Wade, John C Morris, Olivier Rixe, Monica M Mita, Jean-François Dedieu, Claudine Wack, Laurent Kassalow, A Craig Lockhart
ABSTRACT
Cabazitaxel is primarily metabolized by CYP3A. This study evaluated the impact of moderate/strong CYP3A inhibitors [aprepitant (Study Part 2); ketoconazole (Study Part 3)] or strong CYP3A inducers [rifampin (Study Part 4)] on the pharmacokinetics of cabazitaxel. Adult patients received IV cabazitaxel/cisplatin 15/75 mg/m(2) on Day 1 of 3-week cycles (5/75 mg/m(2) in Cycles 1 and 2 of Part 3 to allow a safety margin to the cabazitaxel MTD). Patients received repeated oral doses of aprepitant, ketoconazole or rifampin before/during Cycle 2. Cabazitaxel clearance was the primary endpoint; clearance and area under the plasma concentration-time curve (AUC) were normalized to body surface area and dose, respectively. The PK population included 13 (Part 2), 23 (Part 3) and 21 patients (Part 4). Repeated aprepitant administration did not affect cabazitaxel clearance [geometric mean ratio (GMR) 0.98; 90 % confidence interval (CI) 0.80-1.19]. Repeated ketoconazole administration resulted in 20 % decrease in cabazitaxel clearance (GMR 0.80; 90 % CI 0.55-1.15), associated with 25 % increase in AUC (GMR 1.25; 90 % CI 0.86-1.81). Repeated rifampin administration resulted in 21 % increase in cabazitaxel clearance (GMR 1.21; 90 % CI 0.95-1.53), associated with 17 % decrease in AUC (GMR 0.83; 90 % CI 0.65-1.05). The GMR of AUC0-24 with rifampin administration was 1.09 (90 % CI 0.9-1.33), suggesting that rifampin had a low impact during the initial phases of cabazitaxel elimination. Safety findings were consistent with previous results. Cabazitaxel pharmacokinetics are modified by drugs strongly affecting CYP3A. Co-administration of cabazitaxel with strong CYP3A inhibitors or inducers should be avoided.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Rifampicin, European Pharmacopoeia (EP) Reference Standard
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material