MilliporeSigma
  • Attitudes to mesalamine questionnaire: a novel tool to predict mesalamine nonadherence in patients with IBD.
Attitudes to mesalamine questionnaire: a novel tool to predict mesalamine nonadherence in patients with IBD.
The American journal of gastroenterology (2014-06-11)
Alan C Moss, Yvonne Lillis, Jessica B Edwards George, Niteesh K Choudhry, Anders H Berg, Adam S Cheifetz, Gary Horowitz, Dan A Leffler
ABSTRACT
Poor adherence to mesalamine is common and driven by a combination of lifestyle and behavioral factors, as well as health beliefs. We sought to develop a valid tool to identify barriers to patient adherence and predict those at risk for future nonadherence. A 10-item survey was developed from patient-reported barriers to adherence. The survey was administered to 106 patients with ulcerative colitis who were prescribed mesalamine, and correlated with prospectively collected 12-month pharmacy refills (medication possession ratio (MPR)), urine levels of salicylates, and self-reported adherence (Morisky Medication Adherence Scale (MMAS)-8). From the initial 10-item survey, 8 items correlated highly with the MMAS-8 score at enrollment. Computer-generated randomization produced a derivation cohort of 60 subjects and a validation cohort of 46 subjects to assess the survey items in their ability to predict future adherence. Two items from the patient survey correlated with objective measures of long-term adherence: their belief in the importance of maintenance mesalamine even when in remission and their concerns about side effects. The additive score based on these two items correlated with 12-month MPR in both the derivation and validation cohorts (P<0.05). Scores on these two items were associated with a higher risk of being nonadherent over the subsequent 12 months (relative risk (RR) =2.2, 95% confidence interval=1.5-3.5, P=0.04). The area under the curve for the performance of this 2-item tool was greater than that of the 10-item MMAS-8 score for predicting MPR scores over 12 months (area under the curve 0.7 vs. 0.5). Patients' beliefs about the need for maintenance mesalamine and their concerns about side effects influence their adherence to mesalamine over time. These concerns could easily be raised in practice to identify patients at risk of nonadherence (Clinical Trial number NCT01349504).
Product Description
Supelco
Mesalamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Mesalazine, European Pharmacopoeia (EP) Reference Standard
Mesalazine for system suitability, European Pharmacopoeia (EP) Reference Standard