MilliporeSigma
  • Selective determination of melamine in aqueous medium by molecularly imprinted solid phase extraction.
Selective determination of melamine in aqueous medium by molecularly imprinted solid phase extraction.
Journal of agricultural and food chemistry (2010-10-26)
Manuela Curcio, Francesco Puoci, Giuseppe Cirillo, Francesca Iemma, Umile Gianfranco Spizzirri, Nevio Picci
ABSTRACT
A molecularly imprinted polymer able to recognize melamine in partially aqueous medium was synthesized using methacrylic acid as functional monomer and ethylene glycol dimethacrylate as cross-linking agent. The bound specificity and selectivity of the obtained material were verified by performing binding experiments with melamine and its structural analogue, 2,4,6-trimethoxy-1,3,5-triazine, respectively, using different aqueous binding media. Finally, the ability of MIP to selectively extract melamine from two real samples, a food supplement and a freeze-dried meat sample, was demonstrated.